Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019523.4 | XP_004019572.1 | 4 - 5 | 1243 | 87 | |
XM_012101874.3 | XP_011957264.1 | 4 - 5 | 1487 | 87 | ||
XM_012101875.3 | XP_011957265.1 | 4 - 5 | 1442 | 87 | ||
XM_012101876.3 | XP_011957266.1 | 4 - 5 | 1569 | 87 | ||
XM_027959265.1 | XP_027815066.1 | 5 - 6 | 1610 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |