Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969808.1 | XP_027825609.1 | 1 - 2 | 548 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |