Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973969.1 | XP_027829770.1 | 1 - 2 | 411 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |