Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008427.4 | XP_004008476.1 | 1 - 2 | 372 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |