Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977241.1 | XP_027833042.1 | 7 - 8 | 925 | 63 | |
XM_027977242.1 | XP_027833043.1 | 7 - 8 | 926 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |