Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098988.2 | XP_014954474.2 | 5 - 6 | 1458 | 130 | |
XM_027973993.1 | XP_027829794.1 | 4 - 5 | 2272 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |