Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006863.4 | XP_004006912.1 | 4 - 5 | 396 | 141 | |
XM_012175354.3 | XP_012030744.1 | 4 - 5 | 325 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |