Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975238.1 | XP_027831039.1 | 2 - 3 | 1340 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |