Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966459.1 | XP_027822260.1 | 3 - 4 | 627 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |