Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966791.1 | XP_027822592.1 | 2 - 3 | 1392 | 116 | |
XM_027966795.1 | XP_027822596.1 | 2 - 3 | 1385 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |