Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963616.1 | XP_027819417.1 | 6 - 7 | 1085 | 60 | |
XM_027963617.1 | XP_027819418.1 | 6 - 7 | 1086 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |