Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969506.1 | XP_027825307.1 | 2 - 3 | 1300 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |