Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006199.4 | XP_004006248.3 | 1 - 2 | 178 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |