Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012120268.2 | XP_011975658.2 | 2 - 3 | 1978 | 96 | |
XM_027964393.1 | XP_027820194.1 | 4 - 5 | 1794 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |