Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009867.2 | XP_004009916.2 | 1 - 2 | 125 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |