Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015097.4 | XP_004015146.2 | 2 - 3 | 242 | 88 | |
XM_012190032.3 | XP_012045422.2 | 2 - 3 | 181 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |