Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959276.1 | XP_027815077.1 | 5 - 6 | 1346 | 76 | |
XM_027959277.1 | XP_027815078.1 | 5 - 6 | 3418 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |