Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976143.1 | XP_027831944.1 | 2 - 3 | 391 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |