Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967612.1 | XP_027823413.1 | 4 - 5 | 1802 | 82 | |
XM_027967613.1 | XP_027823414.1 | 4 - 5 | 1848 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |