Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022232.4 | XP_004022281.1 | 13 - 14 | 1620 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |