Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106873.3 | XP_011962263.2 | 2 - 3 | 1894 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |