Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008812.4 | XP_004008861.2 | 3 - 4 | 188 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |