Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011178.4 | XP_004011227.3 | 1 - 2 | 785 | 91 | |
XM_004011179.4 | XP_004011228.3 | 1 - 2 | 785 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |