Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015395.3 | XP_004015444.2 | 3 - 4 | 378 | 72 | |
XM_012094665.3 | XP_011950055.2 | 3 - 4 | 380 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |