Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958994.1 | XP_027814795.1 | 3 - 4 | 1314 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |