Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101110605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186735.3 | XP_012042125.3 | 1 - 2 | 173 | 64 | |
XM_015098674.2 | XP_014954160.2 | 1 - 2 | 173 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |