Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975991.1 | XP_027831792.1 | 2 - 3 | 830 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |