Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099541.2 | XP_014955027.2 | 3 - 4 | 584 | 95 | |
XM_027976531.1 | XP_027832332.1 | 3 - 4 | 584 | 95 |