Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014287.3 | XP_004014336.1 | 4 - 5 | 1523 | 64 | |
XM_012188506.2 | XP_012043896.1 | 4 - 5 | 1033 | 64 | ||
XM_027976728.1 | XP_027832529.1 | 4 - 5 | 1033 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |