Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968587.1 | XP_027824388.1 | 2 - 3 | 299 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |