Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEDDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976128.1 | XP_027831929.1 | 1 | 858 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |