Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023365.3 | XP_004023414.1 | 1 | 818 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |