Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005072.4 | XP_004005121.3 | 5 - 6 | 1196 | 101 | |
XM_012150886.3 | XP_012006276.2 | 4 - 5 | 1077 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |