Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010053.4 | XP_004010102.3 | 2 - 3 | 1042 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |