Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014414.4 | XP_004014463.1 | 3 - 4 | 584 | 102 | |
XM_027976962.1 | XP_027832763.1 | 3 - 4 | 533 | 102 | ||
XM_027976963.1 | XP_027832764.1 | 3 - 4 | 711 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |