Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCAY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969868.1 | XP_027825669.1 | 11 - 12 | 1485 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |