Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020462.4 | XP_004020511.1 | 4 - 5 | 772 | 98 | |
XM_012103453.3 | XP_011958843.1 | 4 - 5 | 807 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |