Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962090.1 | XP_027817891.1 | 2 - 3 | 373 | 110 | |
XM_027962091.1 | XP_027817892.1 | 2 - 3 | 1576 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |