Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976265.1 | XP_027832066.1 | 3 - 4 | 3411 | 103 | |
XM_027976266.1 | XP_027832067.1 | 4 - 5 | 2924 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |