Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962318.1 | XP_027818119.1 | 3 - 4 | 225 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |