Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008724.4 | XP_004008773.1 | 9 - 10 | 1193 | 108 | |
XM_012178314.3 | XP_012033704.1 | 11 - 12 | 1335 | 108 | ||
XM_012178315.3 | XP_012033705.1 | 10 - 11 | 1245 | 108 | ||
XM_012178316.2 | XP_012033706.1 | 9 - 10 | 1037 | 108 | ||
XM_012178319.3 | XP_012033709.1 | 9 - 10 | 930 | 108 | ||
XM_012178324.2 | XP_012033714.1 | 8 - 9 | 911 | 108 | ||
XM_027970041.1 | XP_027825842.1 | 9 - 10 | 1314 | 108 | ||
XM_027970042.1 | XP_027825843.1 | 11 - 12 | 1309 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |