Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010255.4 | XP_004010304.2 | 2 - 3 | 1992 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |