Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019146.4 | XP_004019195.1 | 1 - 2 | 190 | 92 | |
XM_027958613.1 | XP_027814414.1 | 1 - 2 | 325 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |