Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102042.2 | XP_011957432.1 | 11 - 12 | 1457 | 106 | |
XM_012102044.3 | XP_011957434.1 | 11 - 12 | 1472 | 106 | ||
XM_012102045.3 | XP_011957435.1 | 11 - 12 | 1491 | 106 | ||
XM_012102047.2 | XP_011957437.1 | 11 - 12 | 1823 | 106 | ||
XM_012102048.3 | XP_011957438.1 | 10 - 11 | 1225 | 106 | ||
XM_012102049.3 | XP_011957439.1 | 10 - 11 | 1317 | 106 | ||
XM_027960097.1 | XP_027815898.1 | 11 - 12 | 1455 | 106 | ||
XM_027960098.1 | XP_027815899.1 | 11 - 12 | 1452 | 106 | ||
XM_027960099.1 | XP_027815900.1 | 11 - 12 | 1446 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |