Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096241.2 | XP_011951631.1 | 1 | 623 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |