Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970589.1 | XP_027826390.1 | 4 - 5 | 328 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |