Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009827.4 | XP_004009876.2 | 8 - 9 | 2085 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |