Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016999.3 | XP_004017048.2 | 4 - 5 | 503 | 98 | |
XM_012096943.2 | XP_011952333.2 | 4 - 5 | 505 | 98 | ||
XM_012096945.3 | XP_011952335.1 | 3 - 4 | 434 | 98 | ||
XM_027980074.1 | XP_027835875.1 | 5 - 6 | 648 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |