Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975703.1 | XP_027831504.1 | 1 - 2 | 291 | 64 | |
XM_027975704.1 | XP_027831505.1 | 1 - 2 | 291 | 64 | ||
XM_027975705.1 | XP_027831506.1 | 1 - 2 | 291 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |