Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005788.3 | XP_004005837.1 | 3 - 4 | 2307 | 89 | |
XM_027966637.1 | XP_027822438.1 | 3 - 4 | 1306 | 89 | ||
XM_027966638.1 | XP_027822439.1 | 3 - 4 | 2304 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |